Learn More
Thermo Scientific™ Pierce™ Endotoxin-Free Tris Buffer, pH 7.0 (50 mM)
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
Pack of 5
Description
Pierce Endotoxin-Free Tris Buffer can be used to dilute or adjust the pH of samples for endotoxin detection (LAL) assay. For endotoxin testing, the sample pH should be between pH 6-8. The buffer contains <0.005 EU/mL endotoxin.
Specifications
Specifications
| Description | Endotoxin-Free Tris Buffer |
| Concentration | 50 mM |
| For Use With (Application) | For adjusting the pH of the LAL endotoxin test samples |
| For Use With (Equipment) | LAL Endotoxin Assay |
| pH | 7 |
| Quantity | 5 x 10 mL |
| Packaging | Vial(s) |
| Sterility | Sterile |
| Endotoxin Concentration | <0.005 EU/mL |