Learn More
Thermo Scientific™ Pierce™ Mercaptoethylamine-HCl
Product Code. 11816784 Shop All Thermo Scientific Products
Click to view available options
Quantity:
6 x 6 mg
Unit Size:
Pack of 6
Product Code. 11816784
Brand: Thermo Scientific™ 20408
Pure, crystalline 2-aminoethanethiol (2-MEA-HCl, also called cysteamine-HCl), CAS 156-57-0; selectively reduces antibody hinge-region disulfide bonds.
Thermo Scientific™ Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.
M.W. 113.61
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store desiccated at 4°C. Product is shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 6 x 6 mg
Product Type Mercaptoethylamine Hydrochloride
compliance-icons
Product Identifier
  • 2-MERCAPTOETHYLAMINE - HCL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2-MercaptoethylamineHydrochloride