missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Piclamilast

Product Code. 18035192
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18035192

Brand: Tocris Bioscience™ 4525/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PDE4 inhibitor

Potent and selective inhibitor of phosphodiesterase (PDE) 4 (IC50 = 1 nM at PDE4 from human neutrophils). Displays >19,000-fold selectivity over other PDE isoenzymes. Exhibits anti-inflammatory effects in models of allergic inflammation. Also inhibits LTB4 synthesis in human neutrophils (IC50 = 2 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144035-83-6
Molecular Formula C18H18Cl2N2O3

Specifications

Quantity 10 mg
Formula Weight 381.25
Purity >99%
Molecular Weight (g/mol) 381.253
Chemical Name or Material Piclamilast
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.