accessibility menu, dialog, popup

UserName

CAS RN 144035-83-6

H3COOOHNClNCl

CAS RN 144035-83-6

IUPAC Name: 3-(cyclopentyloxy)-N-(3,5-dichloropyridin-4-yl)-4-methoxybenzamide
Synonyms: piclamilast 吡拉米司特
Molecular Weight (g/mol): 381.25
Molecular Formula: C18H18Cl2N2O3
InChi Key: RRRUXBQSQLKHEL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC1CCCC1)C(=O)NC1=C(Cl)C=NC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results