missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Piclamilast

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective inhibitor of phosphodiesterase (PDE) 4 (IC50 = 1 nM at PDE4 from human neutrophils). Displays >19,000-fold selectivity over other PDE isoenzymes. Exhibits anti-inflammatory effects in models of allergic inflammation. Also inhibits LTB4 synthesis in human neutrophils (IC50 = 2 nM).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 381.25 |
| Purity | >99% |
| Molecular Weight (g/mol) | 381.253 |
| Chemical Name or Material | Piclamilast |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction