missing translation for 'onlineSavingsMsg'
Learn More

Abcam Piceatannol, Syk Tyrosine Kinase Inhibitor, >99% Purity, 50 mg

Product Code. 32108599 Shop All Abcam Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32108599 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32108599 Supplier Abcam Supplier No. ab12072250mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent Syk tyrosine kinase inhibitor with anti-inflammatory, antiproliferative, and immunomodulatory effects.

  • Molecular Weight: 244.25 g/mol
  • Purity: >99%
  • Potency: Syk tyrosine kinase inhibitor (Ki = 15 μM at Syk)
  • Biological Activity: Exhibits anti-inflammatory, antiproliferative, and immunomodulatory effects
  • Form: Solid
  • CAS Number: 10083-24-6

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Antioxidant
Buffer 100% Piceatannol
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Target Molecule Syk Tyrosine Kinase
Formulation 100% Piceatannol
Quantity 50 mg
For Use With (Application) Syk tyrosine kinase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.