Learn More
Eurofins DiscoverX PI3 Kinase (p110β/p85α) Human Protein
Description
Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.
GenBank NM_006219 / GenBank XM_043865
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 0.2 mM PMSF, 1 mM benzamidine, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 124 kDa (P110β) and 83.7 kDa (p85α) |
| Quantity | 20 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / Untagged |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥67% |
| Protein | PI3 Kinase (p110β/p85α), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.