Learn More
PI3 Kinase p110 Alpha Mouse anti-Human, Unconjugated, Clone: 1F6A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67071-1-IG-20UL
 View more versions of this product
Product Code. 16829485
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
The PI3 Kinase p110 Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Alpha. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The PI3 Kinase p110 Alpha antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PI3 Kinase p110 Alpha
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Co-immunoprecipitation (Co-IP), Immunohistochemistry
1F6A7
Unconjugated
20 μL
5290
IgG2b