Learn More
PI3 Kinase p110 Alpha Mouse anti-Human, Unconjugated, Clone: 1F6A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67071-1-IG-150UL
Description
The PI3 Kinase p110 Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Alpha. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The PI3 Kinase p110 Alpha antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PI3 Kinase p110 Alpha | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit |
| Co-immunoprecipitation (Co-IP), Immunohistochemistry | |
| 1F6A7 | |
| Unconjugated | |
| 150 μL | |
| 5290 | |
| IgG2b |