missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3-Kinase p110 beta/PIK3CB Antibody (CL9518) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88937CL1
This item is not returnable.
View return policy
Description
PI 3-Kinase p110 beta/PIK3CB Monoclonal antibody specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| PI 3-Kinase p110 beta/PIK3CB | |
| Monoclonal | |
| CoraFluor 1 | |
| P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta | |
| This antibody was developed using a synthetic peptide derived from P42338, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, mTOR Pathway | |
| 5291 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL9518 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction