missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PHT 427
Description
PHT 427 is a dual Akt and PDK1 inhibitor; binds the pleckstrin homology (PH) domain of Akt and PDK1 (Ki values are 2.7 and 5.2 μM, respectively). Inhibits Akt phosphorylation and Akt downstream targets; also induces apoptosis in vitro. Displays synergistic antitumor effect when combined with chemotherapeutics in breast cancer mouse models.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | PHT 427 |
| Molecular Weight (g/mol) | 409.61 |
| Inhibitors | PHT 427 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?