missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHPT1 Antibody (PHPT1/12054), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45069
This item is not returnable.
View return policy
Description
PHPT1 Monoclonal antibody specifically detects PHPT1 in Human samples. It is validated for Immunohistochemistry
Specifications
| PHPT1 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry : 1-2ug/mL | |
| bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
| Recombinant human PHPT1 protein (Uniprot: Q9NRX4). Exact sequence is proprietary. | |
| 100 μg | |
| Lipid and Metabolism | |
| 29085 | |
| Store at -20 to -84°C. Avoid freeze-thaw cycles. | |
| Purified |
| Immunohistochemistry | |
| PHPT1/12054 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction