Learn More
Phosphotungstic Acid Solution, Ph. Eur., Reagecon™
BIOPHARMA
Product Code. 11444719 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444719 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11444719 Supplier Reagecon Supplier No. 1065200
  • ONLINE EXCLUSIVE PRICE
    131.00€
    129.10€ / 100mL
    Save 1.90€
    1% Off
 Restricted Item
Estimated Shipment: 13-11-2026