Specifications
| Packaging | Bottle |
| Analyte or Component Names | P |
| Concentration | 1000 mg/L |
| Type | Mass Spectrometry Standard |
| Element | P |
| Solvent or Matrix | Sulfuric Acid |
| Quantity | 100 mL |