missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A (Tyr694)/STAT5B (Tyr699) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28949-1-AP-100UL
This item is not returnable.
View return policy
Description
The Phospho-STAT5A (Tyr694)/STAT5B (Tyr699) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-STAT5A (Tyr694)/STAT5B (Tyr699). This antibody recognizes human antigen. The Phospho-STAT5A (Tyr694)/STAT5B (Tyr699) antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| STAT5A (Tyr694)/STAT5B (Tyr699) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6776 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| 700 μg/mL | |
| MGF; STAT5 | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human | |
| 92-100 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction