missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RPS6KA1 (Thr359/Ser363) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68041-1-IG-20UL
This item is not returnable.
View return policy
Description
The Phospho-RPS6KA1 (Thr359/Ser363) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-RPS6KA1 (Thr359/Ser363). This antibody recognizes human, mouse antigen. The Phospho-RPS6KA1 (Thr359/Ser363) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| RPS6KA1 (Thr359/Ser363) | |
| Monoclonal | |
| 1000 μg/mL | |
| HU-1; MAPKAPK1A; RSK; RSK1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 80 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2C11D9 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6195 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction