missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RPS6KA1 (Ser380) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
119.00€ - 319.00€
Specifications
| Antigen | RPS6KA1 (Ser380) |
|---|---|
| Concentration | 350 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235563
|
Proteintech
28983-1-AP-20UL |
20 μL |
135.00€ 119.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235528
|
Proteintech
28983-1-AP-100UL |
100 μL |
360.00€ 319.00€ / 100µL Save 41.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-RPS6KA1 (Ser380) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-RPS6KA1 (Ser380). This antibody recognizes human antigen. The Phospho-RPS6KA1 (Ser380) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| RPS6KA1 (Ser380) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| HU-1; MAPKAPK1A; RSK; RSK1 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 350 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 6195 | |
| Peptide | |
| Primary | |
| Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title