missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PDPK1 (Ser241) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29241-1-AP-20UL
This item is not returnable.
View return policy
Description
The Phospho-PDPK1 (Ser241) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-PDPK1 (Ser241). This antibody recognizes human, mouse antigen. The Phospho-PDPK1 (Ser241) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| PDPK1 (Ser241) | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5170 | |
| Store at -20°C. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 200 μg/mL | |
| MGC20087; MGC35290; PDK1; PRO0461 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 60-63 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur