missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p70(S6K) (Thr229) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81592-1-RR-20UL
This item is not returnable.
View return policy
Description
The Phospho-p70(S6K) (Thr229) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-p70(S6K) (Thr229). This antibody recognizes human antigen. The Phospho-p70(S6K) (Thr229) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| p70(S6K) (Thr229) | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| PS6K; S6K; S6K1; STK14A; p70(S6K)-alpha; p70-S6K; p70-alpha | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| 65-85 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 2G19 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 6198 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction