Learn More
Phospho-ERK1/2 (Thr202/Tyr204) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
135.00€ - 360.00€
Specifications
| Antigen | Phospho-ERK1/2 (Thr202/Tyr204) |
|---|---|
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16826235
|
Proteintech
28733-1-AP-20UL |
20 μL |
135.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16816235
|
Proteintech
28733-1-AP-100UL |
100 μL |
360.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-ERK1/2 (Thr202/Tyr204) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-ERK1/2 (Thr202/Tyr204). This antibody recognizes human, mouse, rat antigen. The Phospho-ERK1/2 (Thr202/Tyr204) antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| Phospho-ERK1/2 (Thr202/Tyr204) | |
| Polyclonal | |
| Rabbit | |
| 5595 | |
| Primary |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.