missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Connexin 43/GJA1 (S368) Antibody, R&D Systems™
Description
Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide coupled columns.
Connexin 43/GJA1 Polyclonal specifically detects Connexin 43/GJA1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Guinea Pig, Primate, Zebrafish samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Connexin 43/GJA1 (p ser368) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 dilution |
| Gene Alias | connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJA1, GJAL, HSS, ODD, ODDD, ODOD, SDTY3 |
| Gene Symbols | GJA1 |
| Host Species | Rabbit |
| Immunogen | Phosphopeptide corresponding to amino acid residues surrounding the phospho-S368 of Connexin 43/GJA1 |
| Purification Method | Affinity Purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?