missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Phosphatidylserine Antibody (bavituximab) - Chimeric, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Phosphatidylserine Monoclonal antibody specifically detects Phosphatidylserine in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | Phosphatidylserine |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | bavituximab |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | Q9UG56 |
| Host Species | Human |
| Immunogen | Phosphatidylserine |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?