missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Standard Solution, Ion ECON, Chem-Lab
Description
A phosphate standard solution contains a known concentration of phosphate that can be used as a reference standard in analytical testing, since the final value is known. Standards can be used to establish calibration curves, to determine matrix interferences using standard additions, for control charting, and for troubleshooting and verification purposes.
Specifications
Specifications
| CAS | 7664-38-2 |
| Density | 1.00g/mL |
| Molecular Formula | H3O4P |
| Concentration | 1000 μg/mL of PO43− (±7μg, 20°C) |
| InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
| SMILES | OP(=O)(O)O |
| IUPAC Name | phosphoric acid |
| Molecular Weight (g/mol) | 97.994 |
| PubChem CID | 1004 |
| Quantity | 500 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?