missing translation for 'onlineSavingsMsg'
Learn More

Merck Phosphatase, Alkaline, Calf Intestine, Protein

Product Code. 17669895
Change view
Click to view available options
Quantity:
1 kU
5 kU
Unit Size:
1ku
5ku
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17669895 5 kU 5ku
17659895 1 kU 1ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17669895 Supplier Merck Supplier No. 5245725KU
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Native, alkaline phosphatase from calf intestine. Homodimeric enzyme that hydrolyzes phosphate esters of primary and secondary alcohols, amines, and phenols. Useful in hydrolysis of 5'-terminal phosphates of DNA and RNA. Note: 1KU = 1,000 units.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 5mM MgCl2, 5mM Tris-HCl, 100μM ZnCl2, 50% glycerol, pH 7.0.
Preparation Method Dilute buffer or H2O
Quantity 5 kU
Immunogen Phosphatase, Alkaline
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.