Learn More
Phenylphosphinic acid, 98%
Product Code. 11428047
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428047 500 g 500g
11418047 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11428047 Supplier Thermo Scientific Alfa Aesar Supplier No. A17453.36
 Restricted Item
In Stock
Add to basket
CAS: 1779-48-2 | C6H7O2P | 142.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1779-48-2
Molecular Formula C6H7O2P
Molecular Weight (g/mol) 142.09
MDL Number MFCD00002131
InChI Key MLCHBQKMVKNBOV-UHFFFAOYSA-N
Synonym phenylphosphinic acid, benzenephosphinic acid, phosphinic acid, phenyl, benzenephosphonous acid, phenyl phosphinic acid, mlchbqkmvknbov-uhfffaoysa-n, benzolphosphonigs, aure, benzene phosphinic acid, hydroxyphenylphosphineoxide, acmc-1bt5t
PubChem CID 6327116
IUPAC Name hydroxy-oxo-phenylphosphanium
SMILES OP(=O)C1=CC=CC=C1
Specifications
Melting Point 83°C to 85°C
Quantity 500 g
UN Number UN3261
Beilstein 2802244
Formula Weight 142.1
Percent Purity 98%
Chemical Name or Material Phenylphosphinic acid