accessibility menu, dialog, popup

UserName

OPOHHPh

CAS RN 1779-48-2

IUPAC Name: hydroxy(oxo)phenyl-λ⁵-phosphanylium
Synonyms: hydroxy(oxo)phenyl-λ⁵-phosphanylium
Molecular Weight (g/mol): 141.09
Molecular Formula: C6H6O2P
InChi Key: KNQVWTDLQQGKSV-UHFFFAOYNA-O
SMILES: O[P+](=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results