missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthal Solution, Solstice
Specifications
Specifications
| Chemical Name or Material | Phenolphthaline Solution |
| CAS | 64-17-5, 111-46-6, 78-93-3, 77-09-8 |
| CAS Max % | <10%, ≥1%, <3%, <10%, ≥50%, ≤100% |
| For Use With (Application) | Laboratory Chemicals |
| For Use With (Equipment) | PANPEHA Indicator Paper |
| Physical Form | Liquid |
| Quantity | 5 L |
| Solution Type | Mixture |
| Packaging | Plastic Can |
| UN Number | UN1170 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?