Learn More
14-3-3 eta Antibody, FITC, R&D Systems™
Product Code. 18605065 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18605065 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18605065 Supplier Novus Biologicals Supplier No. NBP299490F
  • ONLINE EXCLUSIVE PRICE
    461.16€
    406.35€ / 0.1mL
    Save 54.81€
    11% Off
 Restricted Item
Estimated Shipment: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
14-3-3 eta Polyclonal specifically detects 14-3-3 eta in Human samples. It is validated for Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunoprecipitation, Western Blot
Product Type Antibody
Product Line Novus