missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenactropinium Chloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Phenactropinium Chloride |
| CAS | 3784-89-2 |
| Analyte or Component Names | Phenactropinium Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H28 N O4 . Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | 8-Azoniabicyclo[3.2.1]octane, 3-[(2-hydroxy-2-phenylacetyl)oxy]-8-methyl-8-(2-oxo-2-phenylethyl)-, chloride (1:1), 8-Azoniabicyclo[3.2.1]octane, 3-[(hydroxyphenylacetyl)oxy]-8-methyl-8-(2-oxo-2-phenylethyl)-, chloride (9CI), 8-Phenacylhomatropinium chloride (6CI,7CI), 8-Phenacylhomatropine chloride, N-Phenacylhomatropine chloride, N-Phenacylhomatropinium chloride, Phenactropinium, Phenactropinium bromide, Phenactropinium chloride, Phenacylhomatropinium chloride, ROQ 432, Trophenium |
| InChI Formula | InChI=1S/C24H28NO4.ClH/c1-25(16-22(26)17-8-4-2-5-9-17)19-12-13-20(25)15-21(14-19)29-24(28)23(27)18-10-6-3-7-11-18;/h2-11,19-21,23,27H,12-16H2,1H3;1H/q+1;/p-1/t19-,20+,21+,23?,25?; |
| Solution Type | Neat |
| SMILES | [Cl-].C[N+]1(CC(=O)c2ccccc2)[C@@H]3CC[C@H]1C[C@H](C3)OC(=O)C(O)c4ccccc4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?