Learn More
pH Singles Calibration Solutions in 20mL Pouches, Oakton™
Product Code. p-2369131
Change view
Click to view available options
pH:
10.00
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH unitSize
11965484 10.00 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11965484 Supplier Thermo Scientific Eutech Supplier No. 01X223103
Economical pouches are accurate and convenient.
The pH Singles are ideal for field calibration—just select the pouches needed and carry them to your test site for on-the-spot calibration and verification. No bottles to carry or clean. Pouches are air-tight for maximum freshness and unquestionable reliability. Packets are labeled with temperature versus pH reference tables. Sold in packs of twenty 20mL pouches.
Certifications
NIST traceable, All solutions are certified traceable to NIST™ standards.
TRUSTED_SUSTAINABILITY
Specifications
pH 10.00
Quantity 20 mL Pouches