Learn More
PGC1a Mouse anti-Human, Unconjugated, Clone: 1C1B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66369-1-IG-150UL
This item is not returnable.
View return policy
Description
The PGC1a antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human PGC1a. This antibody recognizes human, mouse, rat antigen. The PGC1a antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PGC1a | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1C1B2 | |
| Unconjugated | |
| 150 μL | |
| 10891 | |
| IgG1 |