missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGC-1 beta Antibody (6C3F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.35€ - 546.00€
Specifications
| Antigen | PGC-1 beta |
|---|---|
| Clone | 6C3F6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656505
|
Novus Biologicals
NBP2-37249-0.025ml |
0.025 mL |
230.00€ 217.35€ / 0.03mL Save 12.65€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18178017
|
Novus Biologicals
NBP2-37249 |
0.1 mL |
578.00€ 546.00€ / 0.10mL Save 32.00€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGC-1 beta Monoclonal specifically detects PGC-1 beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| PGC-1 beta | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp686C1790, ERRL1, PERCFLJ14284, peroxisome proliferator-activated receptor gamma, coactivator 1 beta, PGC1, PGC1Bgamma, coactivator 1, beta, PPARGC1 | |
| PPARGC1B | |
| IgG1 | |
| Affinity Purified |
| 6C3F6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Breast Cancer, Cancer, Chromatin Research, Diabetes Research, Hypoxia, Lipid and Metabolism, Mitochondrial Markers, Neuroscience, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 133522 | |
| Purified recombinant fragment of human PPARGC1B (AA: 195-414) expressed in E. Coli. | |
| Primary | |
| 113.2 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title