missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX13 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97247DL594
This item is not returnable.
View return policy
Description
PEX13 Polyclonal antibody specifically detects PEX13 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PEX13 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 5194 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| NALD, Peroxin-13, peroxisomal biogenesis factor 13, peroxisomal membrane protein PEX13, peroxisome biogenesis factor 13, ZWS | |
| Produced in rabbits immunized with E. coli-derived Human PEX13 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction