Learn More
Pesticide-Mix 60 10 μg/mL in Acetone, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Atrazine∣Atrazine-desethyl∣Atrazine-desisopropyl∣Propazine∣Sebuthylazine∣Sebuthylazine-desethyl∣Simazine∣Terbuthylazine∣Terbuthylazine-desethyl |
| CAS | 1912-24-9, 6190-65-4, 1007-28-9, 139-40-2, 7286-69-3, 37019-18-4 |
| Analyte or Component Names | Atrazine∣Atrazine-desethyl∣Atrazine-desisopropyl∣Propazine∣Sebuthylazine∣Sebuthylazine-desethyl∣Simazine∣Terbuthylazine∣Terbuthylazine-desethyl |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO/IEC 17025 |
| Solution Type | Mixture |
| Solvent or Matrix | Acetone |
| Industry Type | Environmental Testing |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Atrazine 10 μg/mL∣Atrazine-desethyl 10 μg/mL∣Atrazine-desisopropyl 10 μg/mL∣Propazine 10 μg/mL∣Sebuthylazine 10 μg/mL∣Sebuthylazine-desethyl 10 μg/mL∣Simazine 10 μg/mL∣Terbuthylazine 10 μg/mL∣Terbuthylazine-desethyl 10 μg/mL |