missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 2 Antibody (SAIC-34B-124) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

244.00€ - 578.00€

Specifications

Antigen Peroxiredoxin 2
Clone SAIC-34B-124
Concentration 1 mg/mL
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18675154
View Documents
Novus Biologicals
NBP3-20102-0.025mg
0.025 mg
244.00€
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673115
View Documents
Novus Biologicals
NBP3-20102-0.2mg
0.2 mg
578.00€
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Peroxiredoxin 2 Monoclonal antibody specifically detects Peroxiredoxin 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Specifications

Specifications

Peroxiredoxin 2
1 mg/mL
Monoclonal
Purified
RUO
Human
EC 1.11.1, MGC4104, Natural killer cell-enhancing factor B, natural killer-enhancing factor B, NKEFBNKEF-B, peroxiredoxin 2, peroxiredoxin-2, PRPTDPX1, PRX2, PRXII, thiol-specific antioxidant 1, Thiol-specific antioxidant protein, Thioredoxin peroxidase 1, Thioredoxin-dependent peroxide reductase 1, torin, TPX1, TSAEC 1.11.1.15
Peptide 'LSEDYGVLK', derived from the cellular redox regulator Peroxiredoxin-2, conjugated to KLH.
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
SAIC-34B-124
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry
Unconjugated
Rabbit
Cancer
PBS
7001
IgG κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.