Learn More
Abcam Periodic Acid Schiff (PAS) Stain Kit (Mucin Stain)
Product Code. 32119694 Shop All Abcam Products
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119694 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32119694 Supplier Abcam Supplier No. ab15068030mL
 Restricted Item
Estimated Shipment: 19-11-2026
Stain Orthoperiodic acid in 45m in Cell/Tissue Extracts with Periodic Acid Schiff (PAS) Stain Kit AB150680. Cited in 126 publications.
Stain Orthoperiodic acid in 45m in Cell/Tissue Extracts with Periodic Acid Schiff (PAS) Stain Kit AB150680. Cited in 126 publications.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 30 mL
Regulatory Status RUO
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.