missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perilipin-2/ADFP Rabbit anti-Human, Clone: ADFP/2755R, Novus Biologicals™
Rabbit Monoclonal Antibody
528.00€
Specifications
| Antigen | Perilipin-2/ADFP |
|---|---|
| Clone | ADFP/2755R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
| Perilipin-2/ADFP | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 123 | |
| IgG | |
| Protein A or G purified |
| ADFP/2755R | |
| Western Blot 1 to 2 μg/mL, ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
| Recombinant fragment (around aa 249-376) of human Perilipin-2/ADFP protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title