missing translation for 'onlineSavingsMsg'
Learn More

Melford Peptone, Powder

Product Code. 30018655
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018655

Brand: Melford P202401000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue.

Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.

Total Nitrogen (TN): 15.0-16.0%

Amino Nitrogen (AN): 3.0-3.5%

AN/TN Ratio: 0.21-0.23

Specifications

Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.