Learn More
Melford Peptone, Granulated
Product Code. 30018661
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018661 500 g 500g
30018662 5 kg 5kg
30018660 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30018661 Supplier Melford Supplier No. P20250500.0
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
  • Amino Nitrogen (AN): 2.5-3.5%
  • Loss on Drying: <6.0%
Specifications
Quantity 500 g