missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienylhafnium(IV) trichloride, 98+%

Product Code. 11401670
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11401670

Brand: Thermo Scientific Alfa Aesar H27010.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 75181-08-7 | C10H20Cl3Hf-9 | 425.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75181-08-7
Molecular Formula C10H20Cl3Hf-9
Molecular Weight (g/mol) 425.11
MDL Number MFCD00070461
InChI Key DROXWYIBBCYOSR-UHFFFAOYSA-K
Synonym pentamethylcyclopentadienylhafnium trichloride, acmc-20ajym, hafnium,trichloro 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
PubChem CID 53471594
IUPAC Name carbanide;cyclopenta-1,3-diene;hafnium;trichloride
SMILES [CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Cl-].[Cl-].[Cl-].[Hf]

Specifications

Melting Point 170°C to 172°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 420.08
Percent Purity ≥98%
Chemical Name or Material Pentamethylcyclopentadienylhafnium(IV) trichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.