CAS RN 75181-08-7
CAS RN 75181-08-7
IUPAC Name:
cyclopenta-1,3-dien-1-ide hafnium trichloride pentamethanide
Synonyms:
cyclopenta-1,3-dien-1-ide hafnium trichloride pentamethanide
Molecular Weight (g/mol):
425.11
Molecular Formula:
C10H20Cl3Hf
InChi Key:
DROXWYIBBCYOSR-UHFFFAOYSA-K
SMILES:
[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[Cl-].[Cl-].[Cl-].[Hf].C1C=CC=[C-]1
1
–
1
of
1
results
1
Pentamethylcyclopentadienylhafnium(IV) trichloride, 98+%
CAS: 75181-08-7 Molecular Formula: C10H20Cl3Hf-9 Molecular Weight (g/mol): 425.11 MDL Number: MFCD00070461 InChI Key: DROXWYIBBCYOSR-UHFFFAOYSA-K Synonym: pentamethylcyclopentadienylhafnium trichloride,acmc-20ajym,hafnium,trichloro 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl PubChem CID: 53471594 IUPAC Name: carbanide;cyclopenta-1,3-diene;hafnium;trichloride SMILES: [CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Cl-].[Cl-].[Cl-].[Hf]