Learn More
Pejvakin Antibody, Novus Biologicals™
Product Code. 18725173 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18725173 - -
18297764 0.025 mL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18725173 Supplier Novus Biologicals Supplier No. NB11075015
Rabbit Polyclonal Antibody
Pejvakin Polyclonal antibody specifically detects Pejvakin in Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).