missing translation for 'onlineSavingsMsg'
Learn More

Pejvakin Antibody, Novus Biologicals™

Product Code. 18297764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18297764 0.025 mL 25µL
18725173 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18297764 Supplier Novus Biologicals Supplier No. NB110750150.025ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pejvakin Polyclonal specifically detects Pejvakin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pejvakin
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000
Formulation Whole antisera with No Preservative
Gene Accession No. Q0ZLH3
Gene Alias Autosomal recessive deafness type 59 protein, deafness, autosomal recessive 59, pejvakin, PJVK
Gene Symbols DFNB59
Host Species Rabbit
Immunogen A synthetic peptide from human Pejvakin conjugated to an immunogenic carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494513
Test Specificity Human. The peptide is homologous in mouse.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.