Learn More
Thermo Scientific™ PEEKsil™ Capillary Tubing
Product Code. 10733697 Shop All Thermo Scientific Products
Click to view available options
Description:
0.002 in. ID; 10 cm length
0.002 in. ID; 20 cm length
0.002 in. ID; 50 cm length
0.004 in. ID; 10 cm length
0.004 in. ID; 20 cm length
0.004 in. ID; 50 cm length
0.007 in. ID; 10 cm length
0.007 in. ID; 20 cm length
0.007 in. ID; 50 cm length
Unit Size:
Pack of 2
Pack of 5
Product Code. 10733697
Brand: Thermo Scientific™ 60182501
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Get excellent chemical compatibility and very low carryover using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing.
Get excellent chemical compatibility and very low carryover to improve liquid chromatography (LC) performance using the highly inert Thermo Scientific™ PEEKsil™ Capillary Tubing. Made of precision-bore fused silica tubing coated with a 1/16 in. O.D. PEEK covering, it is capable of withstanding high pressures. PEEKsil tubing is ideal for capillary HPLC and LC-mass spectrometry (MS). The very smooth internal surface ensures excellent flow characteristics and very low carryover. It is compatible with most organic solvents and is usable in most standard chromatography systems.
Recommended Use
  • Tubing is stiff; not recommended for uses requiring that tubing bends
  • Use precut lengths only; tube cutting after receipt may damage tubing
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC, LC/MS
Type Capillary Tubing
Description 0.002 in. ID; 20 cm length
For Use With (Application) HPLC, LC/MS
Diameter (English) Inner 0.002 in.
Diameter (English) Outer 1/16 in.
Diameter (Metric) Inner 0.051 mm
Diameter (Metric) Outer 1.588 mm
Overall Length (English) 7.874 in.
Overall Length (Metric) 20 cm
Material PEEK, Silica