Learn More
Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections
Product Code. 10716163 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cross
Tee
Union
Diameter (English) Inner:
0.01 in.
0.02 in.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Diameter (English) Inner Unit Size
10716163 Union 0.01 in. Each
10422155 Union 0.02 in. Each
10392975 Tee 0.02 in. Each
10149534 Cross 0.02 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10716163 Supplier Thermo Scientific™ Supplier No. P742
Estimated Shipment: 09-11-2026
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections. The material is biocompatible for a wide range of applications.
  • Absolute zero or low dead volume formats
  • Biocompatible
TRUSTED_SUSTAINABILITY
Specifications
Type Union
Diameter (English) Inner 0.01 in.
Description PEEK Union; 0.070 uL Swept Volume
Diameter (Metric) Inner 7.87 mm
For Use With (Equipment) Tubing
Length (Metric) 23.37 mm