Learn More
Thermo Scientific™ PEEK Union Adapter Assembly
Product Code. 15967881 Shop All Thermo Scientific Products
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Product Code. 15967881
Brand: Thermo Scientific™ P74201
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
  • Absolute zero or low dead volume formats
  • Biocompatible
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1 Each
Includes (2) F-300 Fingertight Fittings
Thread Size 10-32
Description Union Adapter Assembly
For Use With For 1/16 in. Tubing O.D.
Material PEEK
Pressure 5000 psi