Learn More
Cytiva PEEK Stop Plug
Product Code. 11779555 Shop All Cytiva Products
Click to view available options
Connections:
Narrow Stop Plug 1/16 in. Male
Stop Plug 1/16 in. Male
Unit Size:
Each
Pack of 5
Product Code. 11779555
Brand: Cytiva 11000355
Enables connection of HiTrap columns to a syringe, laboratory pump or chromatography system. Cytiva PEEK Stop Plugs are used to seal the top of a chromatography column, e.g. HiTrap™ prepacked columns. Fingertight or standard 1/16 in. or standard 5/16 in. male stop plugs available.
The inlet and outlet design of HiTrap columns eliminates the need for column connectors when used with ÄKTAdesign™ chromatography systems. Connection to a syringe, laboratory pump, or chromatography system such as FPLC™ System is also possible using these connectors.
  • Made of durable polyetheretherketone (PEEK)
  • One fingertight stop plug is connected to each HiTrap column at delivery
  • Available in narrow 1/16 in., standard 1/16 in. and standard 5/16 in. male stop plugs
TRUSTED_SUSTAINABILITY
Specifications
Type Stop Plug
Connections Narrow Stop Plug 1/16 in. Male
Quantity 5 Pk.
Material PEEK
For Use With (Equipment) Chromatography Columns