Learn More
Thermo Scientific™ PEEK Fingertight Fittings
Product Code. 10504465 Shop All Thermo Scientific Products
Click to view available options
Type:
Fingertight Fitting
One-Piece PEEK Fingertight Fitting
One-Piece PEEK Long Fingertight Fitting
PEEK Wing Nut w/PEEK Ferrule
Replacement Ferrule
Short Head Port Plug, Black
Short Head Port Plug, Red
Description:
1/16 in.
1/16 in., 0.37 in. Head
1/16 in., 10-32 Coned, Delrin, Black
1/16 in., 10-32 Coned, Delrin, Red
PEEK
PEEK, 1/32 in., 0.25 in. Head
Unit Size:
Each
Pack of 10
Product Code. 10504465
Brand: Thermo Scientific™ M645X
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
TRUSTED_SUSTAINABILITY
Specifications
Type Fingertight Fitting
Description PEEK, 1/32 in., 0.25 in. Head
For Use With (Equipment) LC Tubing
Material PEEK
Quantity 1 Each