Learn More
Thermo Scientific™ PEEK Capillary Tubing
Product Code. 10552265 Shop All Thermo Scientific Products
Click to view available options
Overall Length (Metric):
5 cm
10 cm
20 cm
30 cm
50 cm
100 cm
Diameter (English) Inner:
0.003 in.
0.005 in.
0.007 in.
0.010 in.
0.020 in.
Color:
Blue
Natural
Orange
Red
Yellow
Unit Size:
Each
Product Code. 10552265
Brand: Thermo Scientific™ 3700510
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors.
Use Thermo Scientific™ Rheodyne™ Sample Loops for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.
  • Broad chemical compatibility
  • Biocompatible
  • Easily cut to desired length
  • Appropriate for many HPLC applications
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
TRUSTED_SUSTAINABILITY
Specifications
For Use With Rheodyne HPLC Sample Injectors
Type Capillary Tubing
Color Red
Diameter (English) Inner 0.005 in.
Diameter (Metric) Inner 0.127 mm
Overall Length (English) 3.937 in.
Overall Length (Metric) 10 cm
Material PEEK