missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98546-50ul
This item is not returnable.
View return policy
Description
PDLIM1 Polyclonal antibody specifically detects PDLIM1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PDLIM1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 μL/mg of lysate | |
| CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human PDLIM1 fragment. | |
| 50 μg | |
| Cancer, Endocrinology | |
| 9124 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction